Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECHANICAL ULTRASONIC BATHS
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952516R
Encompass_Preferred
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M5800, Voltage: 120V, Capacity: 9.5L (2.5 gal.), Dimensions Interior: 29 x 24 x 15cm (11.5 x 9.5 x 6 in.), Hertz: 40kHz, Includes: Tank cover, Plastic Housing, Stainless Steel Tank, Timer: 60 min., Warranty: 2 Years, Weight: 14 lb., 6.4kg
Catalog No. NC3721442