Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECHANICAL ULTRASONIC BATHS
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952816R
Encompass_Preferred
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M8800, Voltage: 120V, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Hertz: 40kHz, Includes: Tank drains with valves, Plastic Housing, Stainless Steel Tank
Catalog No. NC3721443