Learn More
BRANSON ULTRASONICS CORPORATION (3P) MECHANICAL ULTRASONIC BATHS
Catalog No. NC3721428
Encompass_Preferred
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M1800, Voltage: 120V, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Hertz: 40kHz, Includes: Tank cover, mechanical timer, Plastic Housing, Stainless Steel Tank, Timer: 60 min., Warranty: 2 Years
Catalog No. NC3721428 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952116R
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M1800, Voltage: 120V, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Hertz: 40kHz, Includes: Tank cover, mechanical timer, Plastic Housing, Stainless Steel Tank, Timer: 60 min., Warranty: 2 Years