Learn More
Grainger Mechanics Gloves Black/Gray 11 PR
Catalog No. 502767454
Encompass_Preferred
The Grainger 16V390 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48 frame size, CCW rotation and features sleeve bearings. This electric motor can be used for a variety of light industrial, commercial and residential applications. With its compact size and 1/3 horsepower, it is well suited for driving fans, blowers, pumps and other machinery requiring up to 1/3 HP.
Catalog No. 50-276-7454 Supplier Grainger Supplier No. 16V390
The Grainger 16V390 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48 frame size, CCW rotation and features sleeve bearings. This electric motor can be used for a variety of light industrial, commercial and residential applications. With its compact size and 1/3 horsepower, it is well suited for driving fans, blowers, pumps and other machinery requiring up to 1/3 HP.