Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Mechanics Tracks - Fastest Path Tracks
GREENER_CHOICESCI_ED

Catalog No. S01415 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01415 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S01415 Supplier Supertek™ Supplier No. PH10579N/5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fastest Path Tracks is a product from the 6 Mechanics Tracks family set. The 3 metal tracks consists of different path lengths sloping differently. Find the path of the shortest time. Understand the significance of momentum by performing collision experiments using balls of different masses.

  • Impactful way to teach and learn
  • Fun for everyone
  • Metal tracks
  • Sturdy plastic supports
  • Superb finished quality
  • Easy to use and manage
  • Comes assembled
  • Activity Guide included
  • This track is a product from the Supertek 6 Mechanics Tracks family set
  • Enhance the learning of physics when teaching with all 6 products within the family of look alike products

Specifications

Grade 5+
Class Size 1 Class
Product Type Mechanics Tracks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.