Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Mechanics Tracks - Set of all 6 Multi-Activity Tracks
GREENER_CHOICESCI_ED

Catalog No. S01417 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01417 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S01417 Supplier Supertek™ Supplier No. PH10579N/1SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Mechanics Tracks family set is comprised of a collection of metal tracks used to demonstrate and learn about the motion and momentum. It helps in order to study the kinetic and potential energy during the motion of rolling bodies.

  • Planes with Different Inclinations
  • Galileos Inclined Plane
  • Path of a Pendulum
  • Identical Track Race
  • Fastest Path Tracks
  • Loop The Loop
  • Enhance the learning of physics when teaching with all 6 of these products within this family of look alike products
  • Impactful way to teach and learn
  • Fun for everyone
  • Metal tracks
  • Sturdy plastic supports
  • Superb finished quality
  • Easy to use and manage
  • Come assembled
  • Activity Guides included
  • Each product in this set is sold seperately

Specifications

Grade 5+
Class Size 1 Class
Product Type Mechanics Tracks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.