Learn More
MSC MECHANIX STRETCH GLOVE
Catalog No. 5000802132
Encompass
These general purpose gloves, model SFF-91, are designed for versatile applications including equipment operation, construction, and maintenance. Constructed with durable synthetic leather and featuring a smooth grip surface, they offer high visibility and are leather-lined for comfort.
  • General purpose design for various applications
  • Durable synthetic leather construction
  • Smooth grip surface for enhanced handling
  • Hook and loop cuff style ensures a secure fit
  • High visibility lemon yellow color for safety
  • Touchscreen capable and machine washable
  • Provides impact protection and cool ventilation
Catalog No. 50-008-02132 Supplier MSC Supplier No. 70708698
These general purpose gloves, model SFF-91, are designed for versatile applications including equipment operation, construction, and maintenance. Constructed with durable synthetic leather and featuring a smooth grip surface, they offer high visibility and are leather-lined for comfort.
  • General purpose design for various applications
  • Durable synthetic leather construction
  • Smooth grip surface for enhanced handling
  • Hook and loop cuff style ensures a secure fit
  • High visibility lemon yellow color for safety
  • Touchscreen capable and machine washable
  • Provides impact protection and cool ventilation