Learn More
MSC Mechanix Wear CXG-L1 Flame Resistant Gloves, Leather, Large, 1 Pair
Catalog No. 5000795104
Encompass
These flame-resistant gloves are crafted with genuine leather and feature a comfortable top layer of fire-retardant stretch-knit material. They include a wide leather cuff designed for easy entry and a secure fit, making them suitable for high-heat situations. These gloves meet or exceed SFI 3.3/1 specifications.
  • Genuine leather construction
  • Comfortable top layer of fire-retardant CarbonX® stretch-knit
  • Genuine leather palm layered with CarbonX
  • Reinforcement panels in high-wear areas
  • Hook and loop closure provides a secure fit
  • Wide leather cuff allows for easy entry
  • Meets or exceeds SFI 3.3/1, EN 407 X2XXXX, and EN 388 2121X standards
  • Provides heat and flame protection
  • Operating temperature range: 30–400 °F
Catalog No. 50-007-95104 Supplier MSC Supplier No. 36081594
These flame-resistant gloves are crafted with genuine leather and feature a comfortable top layer of fire-retardant stretch-knit material. They include a wide leather cuff designed for easy entry and a secure fit, making them suitable for high-heat situations. These gloves meet or exceed SFI 3.3/1 specifications.
  • Genuine leather construction
  • Comfortable top layer of fire-retardant CarbonX® stretch-knit
  • Genuine leather palm layered with CarbonX
  • Reinforcement panels in high-wear areas
  • Hook and loop closure provides a secure fit
  • Wide leather cuff allows for easy entry
  • Meets or exceeds SFI 3.3/1, EN 407 X2XXXX, and EN 388 2121X standards
  • Provides heat and flame protection
  • Operating temperature range: 30–400 °F