Learn More
MSC Mechanix Wear F9-360 Gloves, Durahide, Cut Resistant, 1 ea
Catalog No. 5000796644
Encompass
The Cut Resistant Leather Driver F9-360 work glove offers reliability and versatility for industrial professionals. Featuring Durahide leather technology, these gloves are durable, water-resistant, and breathable, ensuring comfort in damp conditions with no break-in time.
  • Comfortable stretch-elastic cuff for easy on and off
  • High-tensile strength ArmorCore liner provides 360° EN 388 Level F & ANSI A9 cut resistance
  • Top grain Durahide boar leather protects back of hands
  • Kevlar stitching ensures structural integrity and resilience under heat
  • Durahide leather palm provides extreme abrasion resistance
  • Gunn cut finger design repositions vulnerable seams for improved durability
  • No break-in time required
  • Water-resistant and breathable for dry, comfortable hands
Catalog No. 50-007-96644 Supplier MSC Supplier No. 16333809
The Cut Resistant Leather Driver F9-360 work glove offers reliability and versatility for industrial professionals. Featuring Durahide leather technology, these gloves are durable, water-resistant, and breathable, ensuring comfort in damp conditions with no break-in time.
  • Comfortable stretch-elastic cuff for easy on and off
  • High-tensile strength ArmorCore liner provides 360° EN 388 Level F & ANSI A9 cut resistance
  • Top grain Durahide boar leather protects back of hands
  • Kevlar stitching ensures structural integrity and resilience under heat
  • Durahide leather palm provides extreme abrasion resistance
  • Gunn cut finger design repositions vulnerable seams for improved durability
  • No break-in time required
  • Water-resistant and breathable for dry, comfortable hands