Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ FastFit™ Red Mechanics Gloves

Catalog No. 19030018E Shop All Mechanix Wear Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
2X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-030-018E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-030-018E Supplier Mechanix Wear™ Supplier No. MFF02012
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These high-dexterity, synthetic leather gloves are touchscreen compatible to help users stay connected while on the job. FastFit™ gloves offer ideal fit with TrekDry™ evaporative cooling technology to keep hands cool and comfortable.

  • TrekDry™ material is form-fitting to keep working hands cool and comfortable
  • High-dexterity synthetic leather infused with touchscreen technology
  • Stretch-elastic cuffs create a secure fit
  • Reinforced thumb and index finger provide additional durability
  • Anatomically designed two-piece palm design prevents material bunching
  • Pinched fingertip construction improves fingertip strength and durability
  • Machine washable

Specifications

Back Material TrekDry™
Cuff Material Elastic
Coating Material Uncoated
Color Red
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size 2X-Large
For Use With (Application) Maintenance Repairs, Equipment Operation, and DIY Home Improvement
Glove Cuff Style Open
Series FastFit™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.