Learn More
MSC Mechanix Wear General Purpose Gloves, 3X-Large, High Grip, 1 Pair
Catalog No. 5000686930
Encompass
These general purpose gloves are designed for a secure and comfortable fit, featuring a thermo plastic rubber-lined construction. They are suitable for applications requiring high dexterity and protection.
  • Low-profile thermoplastic elastomer (TPR) closure provides a secure fit to the wrist.
  • Lightweight mesh allows cool air to circulate throughout the glove.
  • Ventilated trigger finger.
  • Ventilated finger fourchettes.
  • Micro-fleece sweat wipe.
  • Anatomical stitch darts conform to the natural curvature of the hand to create a precision fit.
  • Expandable flex joints improve overall finger mobility.
  • Ventilated 0.6mm high-dexterity palm provides evaporative cooling.
  • Touchscreen capable palm & fingers.
  • Nylon carrier loops.
  • Machine washable.
Catalog No. 50-006-86930 Supplier MSC Supplier No. 16569972
These general purpose gloves are designed for a secure and comfortable fit, featuring a thermo plastic rubber-lined construction. They are suitable for applications requiring high dexterity and protection.
  • Low-profile thermoplastic elastomer (TPR) closure provides a secure fit to the wrist.
  • Lightweight mesh allows cool air to circulate throughout the glove.
  • Ventilated trigger finger.
  • Ventilated finger fourchettes.
  • Micro-fleece sweat wipe.
  • Anatomical stitch darts conform to the natural curvature of the hand to create a precision fit.
  • Expandable flex joints improve overall finger mobility.
  • Ventilated 0.6mm high-dexterity palm provides evaporative cooling.
  • Touchscreen capable palm & fingers.
  • Nylon carrier loops.
  • Machine washable.