Learn More
MSC Mechanix Wear Gloves, Durahide Leather, A5 Cut, 1 Pair
Catalog No. 5000796606
Encompass
These durable gloves are engineered for protection against cuts, punctures, and abrasions in demanding environments. Constructed from Durahide leather, they offer reliable hand safety and comfort.
  • Cut, puncture, and abrasion-resistant
  • Machine washable for easy maintenance
  • Water-resistant
  • Full-fingered design with padded palm grip
  • Elastic cuff for a secure fit
  • Meets ANSI Cut Level A5, EN388 3X42E, and ANSI Puncture Level 4 standards
Catalog No. 50-007-96606 Supplier MSC Supplier No. 16333916
These durable gloves are engineered for protection against cuts, punctures, and abrasions in demanding environments. Constructed from Durahide leather, they offer reliable hand safety and comfort.
  • Cut, puncture, and abrasion-resistant
  • Machine washable for easy maintenance
  • Water-resistant
  • Full-fingered design with padded palm grip
  • Elastic cuff for a secure fit
  • Meets ANSI Cut Level A5, EN388 3X42E, and ANSI Puncture Level 4 standards