Learn More
MSC Mechanix Wear Gloves, Large, Synthetic, Smooth Grip, 1 Pair
Catalog No. 5000767636
Encompass
These general purpose gloves are designed for various applications including construction, equipment operation, and maintenance. They are synthetic-lined, 9 in. long, and feature a smooth grip surface.
  • Seamless palm delivers higher abrasion and tear resistance
  • Form-fitting material helps keep hands cool and comfortable
  • Thermal Plastic Rubber closure with hook and loop provides a secure fit
  • Reinforcement panels in high wear areas provide added durability
  • Machine washable
Catalog No. 50-007-67636 Supplier MSC Supplier No. 44888386
These general purpose gloves are designed for various applications including construction, equipment operation, and maintenance. They are synthetic-lined, 9 in. long, and feature a smooth grip surface.
  • Seamless palm delivers higher abrasion and tear resistance
  • Form-fitting material helps keep hands cool and comfortable
  • Thermal Plastic Rubber closure with hook and loop provides a secure fit
  • Reinforcement panels in high wear areas provide added durability
  • Machine washable