Learn More
MSC Mechanix Wear Mechanics Gloves, Hi-Viz FastFit, Synthetic Palm, 1 Pair
Catalog No. 5000687145
Encompass
The Mechanix Wear Hi-Viz FastFit® Mechanics Gloves are designed for multipurpose use, featuring a single layer synthetic leather palm for cut-resistance and an orange TrekDry® back for high-visibility. These gloves are touchscreen compatible and include a pull strap for easy donning. They are silicone-free and latex-free.
  • Synthetic leather palm, single layer
  • TrekDry® back material, high-visibility orange
  • Silicone and latex-free
  • Touchscreen compatible
  • EN 388 rating: 2121X
  • ANSI/ISEA puncture level: 1
Catalog No. 50-006-87145 Supplier MSC Supplier No. 03476264
The Mechanix Wear Hi-Viz FastFit® Mechanics Gloves are designed for multipurpose use, featuring a single layer synthetic leather palm for cut-resistance and an orange TrekDry® back for high-visibility. These gloves are touchscreen compatible and include a pull strap for easy donning. They are silicone-free and latex-free.
  • Synthetic leather palm, single layer
  • TrekDry® back material, high-visibility orange
  • Silicone and latex-free
  • Touchscreen compatible
  • EN 388 rating: 2121X
  • ANSI/ISEA puncture level: 1