Learn More
MSC Mechanix Wear NSMG-08 Gloves, Cut A5, P5, Medium, 1 Pair
Catalog No. 5000793875
Encompass
These gloves are designed for protection against cuts, punctures, and abrasions, featuring synthetic leather with a padded palm grip for enhanced utility.
  • Utility style
  • Uncoated material
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 5
  • Synthetic leather and TrekDry back material
  • Full fingered design
  • Padded palm grip
  • Hook & loop cuff style
  • Machine washable and touchscreen capable
Catalog No. 50-007-93875 Supplier MSC Supplier No. 48654933
These gloves are designed for protection against cuts, punctures, and abrasions, featuring synthetic leather with a padded palm grip for enhanced utility.
  • Utility style
  • Uncoated material
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 5
  • Synthetic leather and TrekDry back material
  • Full fingered design
  • Padded palm grip
  • Hook & loop cuff style
  • Machine washable and touchscreen capable