Learn More
MSC Mechanix Wear Safety Glasses, Full-Framed, Fire Mirror Polycarbonate, 1 ea.
Catalog No. 5000788686
Encompass
These safety glasses feature full-framed black polycarbonate frames with fire mirror polycarbonate lenses. Designed for optimal visibility and protection, they include anti-fog and scratch-resistant coatings, adjustable temples, and UV protection.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lenses enhance durability and clarity
  • Adjustable temples provide a comfortable and secure fit
  • UV protection shields eyes from harmful ultraviolet rays
  • Impact-resistant design for enhanced safety
  • Meets ANSI Z87.1 & CSA Z94.3 standards
Catalog No. 50-007-88686 Supplier MSC Supplier No. 23101397
These safety glasses feature full-framed black polycarbonate frames with fire mirror polycarbonate lenses. Designed for optimal visibility and protection, they include anti-fog and scratch-resistant coatings, adjustable temples, and UV protection.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lenses enhance durability and clarity
  • Adjustable temples provide a comfortable and secure fit
  • UV protection shields eyes from harmful ultraviolet rays
  • Impact-resistant design for enhanced safety
  • Meets ANSI Z87.1 & CSA Z94.3 standards