Learn More
MSC Mechanix Wear SpeedKnit M-Pact Gloves, Nitrile-Coated, Hi-Viz, Large, 1 Pair
Catalog No. 5000795912
Encompass
These SpeedKnit M-Pact work gloves are designed to protect hands with functional impact resistance and high abrasion resistance. They feature a 15-gauge nylon knit coated with sandy nitrile, achieving ANSI Level 4 abrasion resistance. Ergonomically designed Thermoplastic Rubber (TPR) is sonic welded for a seamless fit, providing ANSI Level 1 impact protection.
  • High visibility 15-gauge knit nylon construction
  • Comfortable compression cuffs provide a secure fit
  • Anatomical Thermoplastic Rubber (TPR) meets ANSI 138 Level 1 impact standard
  • Rugged sandy nitrile palm coating with textured grip offers high abrasion resistance and dry grip performance
  • Durability performance meets ANSI Level 4 abrasion standard
  • Touchscreen capable
  • REACH Compliant
  • Machine washable and dryable
  • Meets ANSI/ISEA Cut Level 1
Catalog No. 50-007-95912 Supplier MSC Supplier No. 20376463
These SpeedKnit M-Pact work gloves are designed to protect hands with functional impact resistance and high abrasion resistance. They feature a 15-gauge nylon knit coated with sandy nitrile, achieving ANSI Level 4 abrasion resistance. Ergonomically designed Thermoplastic Rubber (TPR) is sonic welded for a seamless fit, providing ANSI Level 1 impact protection.
  • High visibility 15-gauge knit nylon construction
  • Comfortable compression cuffs provide a secure fit
  • Anatomical Thermoplastic Rubber (TPR) meets ANSI 138 Level 1 impact standard
  • Rugged sandy nitrile palm coating with textured grip offers high abrasion resistance and dry grip performance
  • Durability performance meets ANSI Level 4 abrasion standard
  • Touchscreen capable
  • REACH Compliant
  • Machine washable and dryable
  • Meets ANSI/ISEA Cut Level 1