Learn More
MSC Mechanix Wear Tactical Gloves, 2X-Large, Leather, Smooth Grip, 1 Pair
Catalog No. 5000803080
Encompass
These tactical gloves are designed for high-performance applications, featuring a 2X-Large size and a smooth grip. Lined with Thermo Plastic Rubber, they offer a combination of protection and dexterity for various tasks.
  • Low-profile thermoplastic elastomer (TPR) closure provides a secure fit to the wrist.
  • Lightweight Trekdry conforms to the back of the hand to reduce heat buildup and perspiration.
  • Anatomical stitch darts conform to the natural curvature of the hand to create a precision fit.
  • Expandable flex joints improve trigger finger mobility.
  • High-dexterity 0.5 mm Ax-Suede provides precision feel and high dexterity.
  • Nylon carrier loops.
  • Machine washable.
Catalog No. 50-008-03080 Supplier MSC Supplier No. 47449681
These tactical gloves are designed for high-performance applications, featuring a 2X-Large size and a smooth grip. Lined with Thermo Plastic Rubber, they offer a combination of protection and dexterity for various tasks.
  • Low-profile thermoplastic elastomer (TPR) closure provides a secure fit to the wrist.
  • Lightweight Trekdry conforms to the back of the hand to reduce heat buildup and perspiration.
  • Anatomical stitch darts conform to the natural curvature of the hand to create a precision fit.
  • Expandable flex joints improve trigger finger mobility.
  • High-dexterity 0.5 mm Ax-Suede provides precision feel and high dexterity.
  • Nylon carrier loops.
  • Machine washable.