Learn More
MSC Mechanix Wear Tactical Gloves, Leather, X-Large, Smooth Grip, 1 Pair
Catalog No. 5000793826
Encompass
These tactical gloves are designed for shooting sports, law enforcement, and military applications, providing enhanced dexterity and a secure fit. They feature materials that reduce heat buildup and conform to the hand for precision.
  • Low-profile Thermoplastic Elastomer (TPR) closure provides a secure fit to the wrist
  • Lightweight TrekDry material conforms to the back of the hand to reduce heat buildup and perspiration
  • Anatomical stitch darts conform to the natural curvature of the hand to create a precision fit
  • Expandable flex joints improve trigger finger mobility
  • High-dexterity 0.5 mm Ax-Suede provides precision feel
  • Nylon carrier loops
  • Machine washable
  • Meets EN 388 1121X standards
Catalog No. 50-007-93826 Supplier MSC Supplier No. 47449400
These tactical gloves are designed for shooting sports, law enforcement, and military applications, providing enhanced dexterity and a secure fit. They feature materials that reduce heat buildup and conform to the hand for precision.
  • Low-profile Thermoplastic Elastomer (TPR) closure provides a secure fit to the wrist
  • Lightweight TrekDry material conforms to the back of the hand to reduce heat buildup and perspiration
  • Anatomical stitch darts conform to the natural curvature of the hand to create a precision fit
  • Expandable flex joints improve trigger finger mobility
  • High-dexterity 0.5 mm Ax-Suede provides precision feel
  • Nylon carrier loops
  • Machine washable
  • Meets EN 388 1121X standards