Learn More
Medchemexpress LLC HY-A0269 10mg Medchemexpress, Mecillinam CAS:32887-01-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA026910MG
Encompass_Preferred
Medchemexpress, HY-A0269 10mg Mecillinam CAS:32887-01-7 Mecillinam (Amdinocillin), the β-lactam antibiotic, has a broad spectrum of activity against gram-negative organisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1013