Learn More
Apexbio Technology LLC Meclizine 2HCl 1104-22-9 5g
SDP
Catalog No. 5000094249
Encompass_Preferred
Meclizine dihydrochloride (Meclizine 2HCl CAS 1104-22-9) is a small molecule antagonist of histamine H1 receptors commonly investigated for its antiemetic properties In addition to its antihistaminic activity meclizine exhibits antimuscarinic effects and inhibits oxidative phosphorylation It has also been identified as an agonist of the mouse constitutive androstane receptor (mCAR) and demonstrates inverse agonist activity at other signaling pathways These multifunctional pharmacological properties make meclizine 2HCl a useful tool in research exploring receptor-mediated signaling mitochondrial function and metabolism-related diseases
Catalog No. 50-000-94249 Supplier Apexbio Technology LLC Supplier No. B17865000
Meclizine dihydrochloride (Meclizine 2HCl CAS 1104-22-9) is a small molecule antagonist of histamine H1 receptors commonly investigated for its antiemetic properties In addition to its antihistaminic activity meclizine exhibits antimuscarinic effects and inhibits oxidative phosphorylation It has also been identified as an agonist of the mouse constitutive androstane receptor (mCAR) and demonstrates inverse agonist activity at other signaling pathways These multifunctional pharmacological properties make meclizine 2HCl a useful tool in research exploring receptor-mediated signaling mitochondrial function and metabolism-related diseases