Learn More
Medchemexpress LLC Meclizine dihydrochloride | 1104-22-9 | 99.95% | 1 ML
SDP
Catalog No. 5000406300
Encompass_Preferred
Meclizine dihydrochloride is an antihistamine that reversibly inhibits the interaction of histamine at the H1 receptors. As a member of the piperazine class of H1 antagonists, it serves as an effective anti-motion sickness agent. This compound can cross the blood-brain barrier, making it suitable for research into polyQ toxicity disorders such as Huntington's disease. It acts as an agonist ligand for mouse constitutive androstane receptor (CAR) and an inverse agonist for Human CAR. When stored as a solution, it should be kept at -80°C for up to 6 months or -20°C for up to 1 month, under sealed conditions and away from moisture.
  • Reversibly inhibits histamine at H1 receptors
  • Effective anti-motion sickness agent
  • Crosses the blood-brain barrier
  • Used for research of polyQ toxicity disorders
  • Agonist for mouse CAR and inverse agonist for human CAR
Catalog No. 50-004-06300 Supplier Medchemexpress LLC Supplier No. HYB034910MM/1ML
Meclizine dihydrochloride is an antihistamine that reversibly inhibits the interaction of histamine at the H1 receptors. As a member of the piperazine class of H1 antagonists, it serves as an effective anti-motion sickness agent. This compound can cross the blood-brain barrier, making it suitable for research into polyQ toxicity disorders such as Huntington's disease. It acts as an agonist ligand for mouse constitutive androstane receptor (CAR) and an inverse agonist for Human CAR. When stored as a solution, it should be kept at -80°C for up to 6 months or -20°C for up to 1 month, under sealed conditions and away from moisture.
  • Reversibly inhibits histamine at H1 receptors
  • Effective anti-motion sickness agent
  • Crosses the blood-brain barrier
  • Used for research of polyQ toxicity disorders
  • Agonist for mouse CAR and inverse agonist for human CAR