Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meclizine dihydrochloride, MP Biomedicals™

Catalog No. ICN15534125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15534125 25 g
ICN15534105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15534125 Supplier MP Biomedicals, Inc Supplier No. 0215534125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1104-22-9
Molecular Formula C25H29Cl3N2
Molecular Weight (g/mol) 463.871
MDL Number MFCD00058199
InChI Key VCTHNOIYJIXQLV-UHFFFAOYSA-N
Synonym meclizine dihydrochloride, meclozine dihydrochloride, bonamine, meclizine hydrochloride, postafen, vertizine, taizer, v-cline, 1-4-chlorophenyl phenyl methyl-4-3-methylbenzyl piperazine dihydrochloride, duremesin
PubChem CID 64713
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride
SMILES CC1=CC(=CC=C1)CN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 463.9
Physical Form Powder
Chemical Name or Material Meclizine dihydrochloride
EINECSNumber 214-164-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.