Learn More
Cayman Chemical MeclIzInhydrochlorIde 10mg
SDP
Catalog No. 502972121
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 250 nM); reduces ovalbumin-induced mast cell degranulation, edema and leukocyte influx in rat lung from 2.5-30 mg/kg
Catalog No. 50-297-2121 Supplier Cayman Chemical Supplier No. 1461510 MG
A histamine H1 receptor antagonist (Ki = 250 nM); reduces ovalbumin-induced mast cell degranulation, edema and leukocyte influx in rat lung from 2.5-30 mg/kg