Learn More
Cayman Chemical Meclizine (hydrochloride), Purity: ≥98%, CAS: 36236-67-6, 5 mg
SDP
Catalog No. 502972835
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 250 nM); reduces ovalbumin-induced mast cell degranulation, edema and leukocyte influx in rat lung from 2.5-30 mg/kg
Catalog No. 50-297-2835 Supplier Cayman Chemical Supplier No. 146155 MG
A histamine H1 receptor antagonist (Ki = 250 nM); reduces ovalbumin-induced mast cell degranulation, edema and leukocyte influx in rat lung from 2.5-30 mg/kg