Learn More
Medchemexpress LLC HY-B1320 50mg Medchemexpress, Meclofenamate (sodium) CAS:6385-02-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB132050MG
Encompass_Preferred
Medchemexpress, HY-B1320 50mg Meclofenamate (sodium) CAS:6385-02-0 Meclofenamic acid sodium is a nonsteroidal anti-inflammatory drug (NSAID) approved for use in arthritis (osteo and rheumatoid), analgesia (mild to moderate pain), dysmenorrhea, and heavy menstrual blood loss (menorrhagia). Meclofenamic acid sodium is a non-selective gap-junction blocker and a highly selective inhibitor of fat mass and obesity-associated (FTO) enzyme inhibitor[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0147