Learn More
Enzo Life Sciences Meclofenamate sodium (5g). CAS: 6385-02-0
SDP
Supplier:  Enzo Life Sciences BMLEI1630005
Encompass_Preferred
Dual 5-lipoxygenase (IC50=46.8 µM) and cyclooxygenase I and II inhibitor (IC50=0.6 µM). Clinically useful NSAID. Purity: ≥98% (HPLC). Solubility: Soluble in water (15mg/ml slightly cloudy). Long Term Storage: Ambient.
Catalog No. 50-201-0581