Learn More
Apexbio Technology LLC Meclofenamate Sodium 6385-02-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089759
Encompass_Preferred
Meclofenamate Sodium (CAS 6385-02-0) is a non-steroidal anti-inflammatory drug (NSAID) that acts by inhibiting cyclooxygenase (COX) enzymes thereby modulating prostaglandin synthesis and attenuating inflammatory signaling Preclinical studies have indicated that Meclofenamate Sodium selectively influences targets involved in lipid metabolism suggesting an expanded role beyond general inflammation suppression Due to its dual inhibitory profile this compound is utilized in biomedical research investigating mechanisms of inflammation as well as in studies focused on metabolic disorders and oncology
Catalog No. 50-000-89759 Supplier Apexbio Technology LLC Supplier No. B47965.1
Meclofenamate Sodium (CAS 6385-02-0) is a non-steroidal anti-inflammatory drug (NSAID) that acts by inhibiting cyclooxygenase (COX) enzymes thereby modulating prostaglandin synthesis and attenuating inflammatory signaling Preclinical studies have indicated that Meclofenamate Sodium selectively influences targets involved in lipid metabolism suggesting an expanded role beyond general inflammation suppression Due to its dual inhibitory profile this compound is utilized in biomedical research investigating mechanisms of inflammation as well as in studies focused on metabolic disorders and oncology