Learn More
Selleck Chemical LLC Meclofenamate Sodium S4295-50mg
SDP
Catalog No. 503110457
Encompass_Preferred
Meclofenamate Sodium(CI 583) is a dual COX-1/COX-2 inhibitor with IC50 of 40 nM and 50 nM respectively used in the treatment of joint muscular pain arthritis and dysmenorrhea Meclofenamate Sodium is a non-selective gap-junction blocker and a highly selective inhibitor of fat mass and obesity-associated (FTO) enzyme inhibitor
Catalog No. 50-311-0457 Supplier Selleck Chemical LLC Supplier No. S429550MG
Add to Cart
Add to Cart
Meclofenamate Sodium(CI 583) is a dual COX-1/COX-2 inhibitor with IC50 of 40 nM and 50 nM respectively used in the treatment of joint muscular pain arthritis and dysmenorrhea Meclofenamate Sodium is a non-selective gap-junction blocker and a highly selective inhibitor of fat mass and obesity-associated (FTO) enzyme inhibitor