Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meclofenamic acid 1mg | 644-62-2 | C14H11Cl2NO2 | 1 MG
SDP

Catalog No. 5000228534
Encompass_Preferred

Meclofenamic acid is a non-steroidal anti-inflammatory compound commonly used in research as a selective inhibitor of the FTO demethylase. It is supplied as a solid research reagent, typically packaged in small quantities (e.g., 1 mg) for biochemical, cellular, and epigenetic studies.

  • Non-steroidal anti-inflammatory compound and selective FTO inhibitor.
  • Chemical formula C14H11Cl2NO2 and molecular weight 296.15 g/mol.
  • Supplied as a powder suitable for biochemical and cell-based assays.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Suitable for assay development, target validation, and epigenetic studies.

Catalog No. 50-002-28534 Supplier Medchemexpress LLC Supplier No. HY1172751MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Meclofenamic acid is a non-steroidal anti-inflammatory compound commonly used in research as a selective inhibitor of the FTO demethylase. It is supplied as a solid research reagent, typically packaged in small quantities (e.g., 1 mg) for biochemical, cellular, and epigenetic studies.

  • Non-steroidal anti-inflammatory compound and selective FTO inhibitor.
  • Chemical formula C14H11Cl2NO2 and molecular weight 296.15 g/mol.
  • Supplied as a powder suitable for biochemical and cell-based assays.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Suitable for assay development, target validation, and epigenetic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.