Learn More
Selleck Chemical LLC Meclofenoxate (Centrophenoxine) HCl S4280-50mg
SDP
Catalog No. 503108935
Encompass_Preferred
Meclofenoxate (Centrophenoxine) HCl is an anti-aging drug used to treat the symptoms of senile dementia and Alzheimer s disease and also inhibits the activity of cholinephosphotransferase
Catalog No. 50-310-8935 Supplier Selleck Chemical LLC Supplier No. S428050MG
Meclofenoxate (Centrophenoxine) HCl is an anti-aging drug used to treat the symptoms of senile dementia and Alzheimer s disease and also inhibits the activity of cholinephosphotransferase