Learn More
Apexbio Technology LLC Meclofenoxate hydrochloride 3685-84-5 200mg
SDP
Catalog No. 5000088224
Encompass_Preferred
Meclofenoxate hydrochloride (CAS 3685-84-5) is a synthetic compound formed from dimethylethanolamine (DMAE) and 4-chlorophenoxyacetic acid (pCPA) It is understood to influence cholinergic neurotransmission likely by increasing acetylcholine levels in the central nervous system through metabolic conversion of DMAE Preclinical studies have demonstrated that meclofenoxate hydrochloride enhances memory retention and supports various aspects of cognitive function This molecule is widely utilized in neurological and cognitive research to explore mechanisms underlying neuroprotection memory enhancement and age-related cognitive decline
Catalog No. 50-000-88224 Supplier Apexbio Technology LLC Supplier No. B1160200
Meclofenoxate hydrochloride (CAS 3685-84-5) is a synthetic compound formed from dimethylethanolamine (DMAE) and 4-chlorophenoxyacetic acid (pCPA) It is understood to influence cholinergic neurotransmission likely by increasing acetylcholine levels in the central nervous system through metabolic conversion of DMAE Preclinical studies have demonstrated that meclofenoxate hydrochloride enhances memory retention and supports various aspects of cognitive function This molecule is widely utilized in neurological and cognitive research to explore mechanisms underlying neuroprotection memory enhancement and age-related cognitive decline