Learn More
Medchemexpress LLC Meclofenoxate hydrochloride | 3685-84-5 | 98.3% | 294.17 | 1 ML
SDP
Catalog No. 5000386711
Encompass_Preferred
Meclofenoxate hydrochloride, also known as Centrophenoxine hydrochloride, is an ester synthesized from DMAE and pCPA. This compound is capable of crossing the blood-brain barrier and can stimulate memory and enhance cognition. This product offers several benefits including:
  • Significantly protects mitochondrial function in primary neurons.
  • Restores mitochondrial morphology and reverses induced mitochondrial damage.
  • Regulates redox balance.
  • Improves behavioral abnormalities and depression-like symptoms in Parkinson's disease models.
  • Reduces anhedonia and motor deficits in behavioral tests.
Catalog No. 50-003-86711 Supplier Medchemexpress LLC Supplier No. HY1755510MM/1ML
Meclofenoxate hydrochloride, also known as Centrophenoxine hydrochloride, is an ester synthesized from DMAE and pCPA. This compound is capable of crossing the blood-brain barrier and can stimulate memory and enhance cognition. This product offers several benefits including:
  • Significantly protects mitochondrial function in primary neurons.
  • Restores mitochondrial morphology and reverses induced mitochondrial damage.
  • Regulates redox balance.
  • Improves behavioral abnormalities and depression-like symptoms in Parkinson's disease models.
  • Reduces anhedonia and motor deficits in behavioral tests.