Learn More
Cayman Chemical Meclofenoxate (hydrochloride), Purity: ≥95%, CAS: 3685-84-5, 10 g
SDP
Catalog No. 502940938
Encompass_Preferred
A nootropic agent; inhibits norepinephrine reuptake by isolated rat striatal synaptosomes (IC50 = 0.5 mM) and 5-HT reuptake by isolated rat cortical synaptosomes (IC50 = 2.7 mM); increases survival of dopaminergic neurons in a C. elegans psd-1 knockdown model of Parkinson's disease; reduces rotenone-induced depletion of dopamine and GSH, lipid peroxidation, production of NO, and cortical and cerebral damage, as well as enhances activity of catalase and superoxide dismutase in a rat model of Parkinson's disease at 100 mg/kg; increases synaptic surface density and reduces age-induced decreases in synaptic numerical density in aged rats at 100 mg/kg; reduces neuronal lipofuscin pigment in the cerebral cortex and hippocampus and improves learning and memory in the T-maze in aged mice
Catalog No. 50-294-0938 Supplier Cayman Chemical Supplier No. 2828310 G
A nootropic agent; inhibits norepinephrine reuptake by isolated rat striatal synaptosomes (IC50 = 0.5 mM) and 5-HT reuptake by isolated rat cortical synaptosomes (IC50 = 2.7 mM); increases survival of dopaminergic neurons in a C. elegans psd-1 knockdown model of Parkinson's disease; reduces rotenone-induced depletion of dopamine and GSH, lipid peroxidation, production of NO, and cortical and cerebral damage, as well as enhances activity of catalase and superoxide dismutase in a rat model of Parkinson's disease at 100 mg/kg; increases synaptic surface density and reduces age-induced decreases in synaptic numerical density in aged rats at 100 mg/kg; reduces neuronal lipofuscin pigment in the cerebral cortex and hippocampus and improves learning and memory in the T-maze in aged mice