Learn More
MSC Made In USA Forklift Drum Caddy, Steel, 55 Gal., 1,000 lbs. Capacity, 1 ea.
Catalog No. 5000512033
Encompass
This forklift drum caddy is designed for the efficient transport, tilting, and dispensing of 55 gal. steel drums. Constructed from heavy-duty steel, the unit attaches to forklift tines to provide a secure and stable way to handle industrial drums in warehouse or manufacturing environments.
  • Features heavy-duty steel construction
  • Compatible with 55 gal. steel drums with 22 in. to 22 1/2 in. diameters
  • Includes 5 in. x 3 in. fork stirrups for mounting to forklift tines
  • Capacity of 1,000 lbs.
  • Secures drums with a steel band featuring an over-center latch
  • Enables drum tilting and dispensing functionality
Catalog No. 50-005-12033 Supplier MSC Supplier No. 89418008
This forklift drum caddy is designed for the efficient transport, tilting, and dispensing of 55 gal. steel drums. Constructed from heavy-duty steel, the unit attaches to forklift tines to provide a secure and stable way to handle industrial drums in warehouse or manufacturing environments.
  • Features heavy-duty steel construction
  • Compatible with 55 gal. steel drums with 22 in. to 22 1/2 in. diameters
  • Includes 5 in. x 3 in. fork stirrups for mounting to forklift tines
  • Capacity of 1,000 lbs.
  • Secures drums with a steel band featuring an over-center latch
  • Enables drum tilting and dispensing functionality