Learn More
Medchemexpress LLC Meconine | 569-31-3 | MFCD00219354 | 98.9% | 194.19 g/mol | C10H10O4 | 200 MG
SDP
Catalog No. 5000300721
Encompass_Preferred
Meconine is an endogenous metabolite of noscapine used as a research reagent and forensic marker for detecting illicit opiate use. It is supplied as a solid and as DMSO solutions for analytical and biological studies. CAS 569-31-3; molecular formula C10H10O4; molecular weight 194.19 g/mol.
  • High purity: 98.9% suitable for analytical applications.
  • Supplied as solid and 10 mM solutions in DMSO for ready use or reconstitution.
  • Useful as a biochemical marker to detect illicit opiate use in biological samples.
  • Available in small pack sizes (100 mg, 200 mg) and larger quantities on request.
  • Documentation includes a safety data sheet with handling and storage recommendations.
Catalog No. 50-003-00721 Supplier Medchemexpress LLC Supplier No. HYW019151200MG
Meconine is an endogenous metabolite of noscapine used as a research reagent and forensic marker for detecting illicit opiate use. It is supplied as a solid and as DMSO solutions for analytical and biological studies. CAS 569-31-3; molecular formula C10H10O4; molecular weight 194.19 g/mol.
  • High purity: 98.9% suitable for analytical applications.
  • Supplied as solid and 10 mM solutions in DMSO for ready use or reconstitution.
  • Useful as a biochemical marker to detect illicit opiate use in biological samples.
  • Available in small pack sizes (100 mg, 200 mg) and larger quantities on request.
  • Documentation includes a safety data sheet with handling and storage recommendations.