Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mecoprop | 93-65-2 | MFCD00002648 | 99.8% | 214.65 g/mol | C10H11ClO3 | 25 G
SDP

Catalog No. 5000307714
Encompass_Preferred

Mecoprop is an analytical standard of the herbicide mecoprop supplied as a solid for research and analytical applications. Documentation including a certificate of analysis and safety data sheet is provided to support laboratory use.

  • High purity analytical standard (99.8%).
  • Chemical formula C10H11ClO3; molecular weight 214.65 g/mol.
  • Supplied as a 25 g solid suitable for laboratory reference and analytical assays.
  • Intended for research and analytical applications; not for human therapeutic use.
  • COA and SDS available for quality and safety information.
  • Recommended storage: powder at -20°C for long-term, 4°C for shorter term; in solvent store at -80°C for extended storage.

Catalog No. 50-003-07714 Supplier Medchemexpress LLC Supplier No. HY11812625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mecoprop is an analytical standard of the herbicide mecoprop supplied as a solid for research and analytical applications. Documentation including a certificate of analysis and safety data sheet is provided to support laboratory use.

  • High purity analytical standard (99.8%).
  • Chemical formula C10H11ClO3; molecular weight 214.65 g/mol.
  • Supplied as a 25 g solid suitable for laboratory reference and analytical assays.
  • Intended for research and analytical applications; not for human therapeutic use.
  • COA and SDS available for quality and safety information.
  • Recommended storage: powder at -20°C for long-term, 4°C for shorter term; in solvent store at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.