Learn More
Aviva Systems Biology Corp MECP2 Antibody - N-terminal region (ARP43584_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43584_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MECP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MECP2. Alternate names: AUTSX3, DKFZp686A24160, MRX16, MRX79, PPMX, RTS, RTT, RS, MRXSL, MRXS13ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-58578