Learn More
Aviva Systems Biology Corp MECP2 Antibody - middle region (ARP38539_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38539_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MECP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MECP2. Alternate names: AUTSX3, DKFZp686A24160, MRX16, MRX79, MRXS13, MRXSL, PPMX, RTS, RTT, RSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56693