Learn More
Strem, An Ascensus Company CAS# [94442-22-5]. 10g. (Trimethyl)methylcyclopentadienylplatinum(IV), 99% (99.999%-Pt) PURATREM.
SDP
Supplier:  Strem, An Ascensus Company 98135010G
Encompass
CAS# [94442-22-5]. 10g. (Trimethyl)methylcyclopentadienylplatinum(IV), 99% (99.999%-Pt) PURATREM. . Molecular Weight: 319.32. Molecular Formula: (CH3)3(CH3C5H4)Pt. Color/form: off-white pwdr. Strem# 98-1350. www.strem.com/catalog/v3/98-1350/
Catalog No. 50-197-9371