Learn More
Strem, An Ascensus Company CAS# [94442-22-5]. 10g. (Trimethyl)methylcyclopentadienylplatinum(IV), 99%, 78-1350, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00079665
SDP
Supplier:  Strem, An Ascensus Company 98402410G
Encompass
CAS# 94442-22-5. 10g. (Trimethyl)methylcyclopentadienylplatinum(IV), 99%, 78-1350, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00079665. Molecular Weight: 319.32. Molecular Formula: (CH3)3(CH3C5H4)Pt. Color/form: off-white pwdr. Strem# 98-4024. www.strem.com/catalog/v3/98-4024/
Catalog No. 50-197-9337