Learn More
Aviva Systems Biology Corp MECR Antibody - C-terminal region (ARP61845_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MECR. It was validated on Western Blot. . Alternate names: CGI-63, FASN2B, NRBF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-485