Learn More
Medchemexpress LLC MeCY5-NHS ester triethylamine | 00-00-0 | 96.2% | 777.94 g/mol | C36H40N3KO10S2 | 5 MG
SDP
Catalog No. 5000221819
Encompass_Preferred
MeCY5-NHS ester (potassium) is an amine-reactive Cy5-family fluorescent dye supplied as the NHS-ester potassium salt for labeling proteins, peptides, and nucleic acids in fluorescence assays and imaging.
  • Reactive N-hydroxysuccinimide (NHS) ester for covalent labeling of primary amines.
  • Cy5-family near-infrared fluorescence, approx. 649/670 nm excitation/emission.
  • Potassium salt form with improved water solubility for aqueous labeling.
  • Supplied with analytical documentation, including SDS, COA, 1H NMR, and LC-MS.
  • Available in lab-scale quantities suitable for conjugation experiments.
Catalog No. 50-002-21819 Supplier Medchemexpress LLC Supplier No. HY135413B5MG
MeCY5-NHS ester (potassium) is an amine-reactive Cy5-family fluorescent dye supplied as the NHS-ester potassium salt for labeling proteins, peptides, and nucleic acids in fluorescence assays and imaging.
  • Reactive N-hydroxysuccinimide (NHS) ester for covalent labeling of primary amines.
  • Cy5-family near-infrared fluorescence, approx. 649/670 nm excitation/emission.
  • Potassium salt form with improved water solubility for aqueous labeling.
  • Supplied with analytical documentation, including SDS, COA, 1H NMR, and LC-MS.
  • Available in lab-scale quantities suitable for conjugation experiments.