Learn More
CELLTREAT Scientific Products 30mL Media Bottle, Square, PETG, Individually Wrapped, Sterile
SDP
Catalog No. NC2545538
Encompass_Preferred
Lightweight plastic material is easier and safer to handle than glass Square media bottle shape conserves space in refrigerated storage areas PET bottles feature clearly printed graduations and defined marking area PETG bottles feature molded graduations All bottles are sterile, and available individually wrapped or tray packed Both PET and PETG materials reduce gas permeability and improve solution stability compared to polystyrene Operating temperature range of –80°C – 60°C
Catalog No. NC2545538 Supplier CELLTREAT Scientific Products Supplier No. 230750
Lightweight plastic material is easier and safer to handle than glass Square media bottle shape conserves space in refrigerated storage areas PET bottles feature clearly printed graduations and defined marking area PETG bottles feature molded graduations All bottles are sterile, and available individually wrapped or tray packed Both PET and PETG materials reduce gas permeability and improve solution stability compared to polystyrene Operating temperature range of –80°C – 60°C