Learn More
CELLTREAT Scientific Products 125mL Media Bottle, Square, PETG, Tamper Evident, Bulk, Sterile
SDP
Catalog No. NC3377725
Encompass_Preferred
Lightweight plastic material is easier and safer to handle than glass Square media bottle shape conserves space in refrigerated storage areas PET bottles feature clearly printed graduations and defined marking area PETG bottles feature molded graduations All bottles are sterile, and available individually wrapped or tray packedTray packed bottles available with tamper evident shrink band option Both PET and PETG materials reduce gas permeability and improve solution stability compared to polystyrene Operating temperature range of –80°C – 60°C
Catalog No. NC3377725 Supplier CELLTREAT Scientific Products Supplier No. 230762TE
Lightweight plastic material is easier and safer to handle than glass Square media bottle shape conserves space in refrigerated storage areas PET bottles feature clearly printed graduations and defined marking area PETG bottles feature molded graduations All bottles are sterile, and available individually wrapped or tray packedTray packed bottles available with tamper evident shrink band option Both PET and PETG materials reduce gas permeability and improve solution stability compared to polystyrene Operating temperature range of –80°C – 60°C