Learn More
Electron Microscopy Sciences Bench Top Medium Dispensing Bin
SDP
Catalog No. 5030599
Encompass_Preferred
Bins are made from 1/4" thick PVC. They are chemical resistant and easy to clean. Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Footprint: 12 x 9"
Height: 6-3/8"
Catalog No. 50-305-99 Supplier Electron Microscopy Sciences Supplier No. 6438812
Bins are made from 1/4" thick PVC. They are chemical resistant and easy to clean. Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Footprint: 12 x 9"
Height: 6-3/8"