Learn More
Electron Microscopy Sciences Anti-Vibration Platform Medium Frame
SDP
Catalog No. 5034459
Encompass_Preferred
Hard grey epoxy powder finish frame that bolts directly to platform. Supplied with four leveling feet providing 3 cm of adjustment. Frame height 71cm, going to a working height of 78cm.
Catalog No. 50-344-59 Supplier Electron Microscopy Sciences Supplier No. 6500
Hard grey epoxy powder finish frame that bolts directly to platform. Supplied with four leveling feet providing 3 cm of adjustment. Frame height 71cm, going to a working height of 78cm.