Learn More
Genesee Scientific Corporation MED ICE PAN,YELLOW 4L W/O LID
SDP
Catalog No. NC2107320
Encompass
4 Liters1 Ice Pan/UnitBrand: Genesee ScientificExcellent insulation, stackable, lightweight and leak-proof designSuperior durability - no sweat, no rust, no deformation and virtually unbreakableChemical resistance - can withstand dry ice, chemical solvents and liquid nitrogenSamples will remain frozen for over 12 hoursRecyclable ethyl-vinyl (EVA) material offers a non-toxic alternative to conventional ice pans and bucketsDimensions (L x W x H): 31.2 x 22.4 x 11.8cm
This item is not returnable. View return policy
Catalog No. NC2107320 Supplier Genesee Scientific Corporation Supplier No. 18302Y
4 Liters1 Ice Pan/UnitBrand: Genesee ScientificExcellent insulation, stackable, lightweight and leak-proof designSuperior durability - no sweat, no rust, no deformation and virtually unbreakableChemical resistance - can withstand dry ice, chemical solvents and liquid nitrogenSamples will remain frozen for over 12 hoursRecyclable ethyl-vinyl (EVA) material offers a non-toxic alternative to conventional ice pans and bucketsDimensions (L x W x H): 31.2 x 22.4 x 11.8cm