Learn More
Andwin Scientific Flame-Resistant Protex Anti-Static String-Knit Gloves, Medium
SDP
Catalog No. NC1540530
Encompass_Preferred
Flame resistant, Protex gloves will not drip, melt or adhere to the skin if exposed to heat or direct flame. These gloves offer great insulation with a thin, seamless, and soft fit. Knit shells are made using THT (True Hand Technology) for a great fit and reduced hand fatigue. Machine washable and quick drying. To limit the risk of allergic reactions, all of our Canadian-made gloves are now crafted with latex-free materials.
This item is not returnable. View return policy
Catalog No. NC1540530 Supplier Andwin Scientific Supplier No. S13FRTMED
Flame resistant, Protex gloves will not drip, melt or adhere to the skin if exposed to heat or direct flame. These gloves offer great insulation with a thin, seamless, and soft fit. Knit shells are made using THT (True Hand Technology) for a great fit and reduced hand fatigue. Machine washable and quick drying. To limit the risk of allergic reactions, all of our Canadian-made gloves are now crafted with latex-free materials.