Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD AFS49ML Performance Medical-Laboratory Freezer 49 Cu. Ft. with 2 Solid Door and casters
SDP
Supplier:  Felix Storch Inc AFS49ML
Encompass_Preferred
ACCUCOLD AFS49ML Performance Medical-Laboratory Freezer 49 Cu. Ft. with 2 Solid Door with Advanced Temperature Control, automatic defrost and forced air cooling. Adjustable operating control range of -25C to -10C with Intelligent Microprocessor Digital Controller designed to provide +/- 2C variation excluding defrost. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Equipped with a fully featured interior for convenient storage. LED lighting, adjustable shelves, and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/AFS49ML
Catalog No. 50-195-9427
Only null left
Add to Cart
Request a Quote